Merck
Indirect short- and long-term effects of aboveground invertebrate and vertebrate herbivores on soil microarthropod communities.
PloS one (2015-03-05)
Martijn L Vandegehuchte, Ursina Raschein, Martin Schütz, Dariusz J Gwiazdowicz, Anita C Risch
RESUMEN
Recognition is growing that besides ungulates, small vertebrate and invertebrate herbivores are important drivers of grassland functioning. Even though soil microarthropods play key roles in several soil processes, effects of herbivores-especially those of smaller body size-on their communities are not well understood. Therefore, we progressively excluded large, medium and small vertebrate and invertebrate herbivores for three growing seasons using size-selective fences in two vegetation types in subalpine grasslands; short-grass and tall-grass vegetation generated by high and low historical levels of ungulate grazing. Herbivore exclusions generally had few effects on microarthropod communities, but exclusion of all herbivore groups resulted in decreased total springtail and Poduromorpha richness compared with exclusion of only ungulates and medium-sized mammals, regardless of vegetation type. The tall-grass vegetation had a higher total springtail richness and mesostigmatid mite abundance than the short-grass vegetation and a different oribatid mite community composition. Although several biotic and abiotic variables differed between the exclusion treatments and vegetation types, effects on soil microarthropods were best explained by differences in nutrient and fibre content of the previous year's vegetation, a proxy for litter quality, and to a lesser extent soil temperature. After three growing seasons, smaller herbivores had a stronger impact on these functionally important soil microarthropod communities than large herbivores. Over longer time-scales, however, large grazers created two different vegetation types and thereby influenced microarthropod communities bottom-up, e.g. by altering resource quality. Hence, both short- and long-term consequences of herbivory affected the structure of the soil microarthropod community.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad