Merck
Insulins, leptin and feeding in a population of Peromyscus leucopus (white-footed mouse) with variable fertility.
Hormones and behavior (2014-03-04)
Jordan T White, Cori L DeSanto, Connie Gibbons, Casey K Lardner, Andrew Panakos, Salehin Rais, Kathy Sharp, Shannon D Sullivan, Wendy Tidhar, Leanne Wright, David Berrigan, Paul D Heideman
RESUMEN
This article is part of a Special Issue "Energy Balance". Natural populations display a variety of reproductive responses to environmental cues, but the underlying physiology that causes these responses is largely unknown. This study tested the hypothesis that heritable variation in reproductive traits can be described by heritable variation in concentrations of hormones critical to both energy balance and reproduction. To test this hypothesis, we used mouse lines derived from a wild population and selectively bred for response to short day photoperiod. Reproductive and metabolic traits of Peromyscus leucopus display heritable variation when held in short photoperiods typical of winter. Our two lines of mice have phenotypes spanning the full range of variation observed in nature in winter. We tested male and female mice for heritable variation in fasted serum concentrations of three hormones involved in energetic regulation: leptin, insulin-like growth factor 1 (IGF-1) and insulin, as well as the effects of exogenous leptin and a high energy diet on reproductive maturation. Exogenous leptin decreased food intake, but protected males from the reduction in testis mass caused by equivalent food restriction in pair-fed, saline-infused controls. A high energy diet resulted in calorie adjustment by the mice, and failed to alter reproductive phenotype. Concentrations of the three hormones did not differ significantly between selection lines but had correlations with measures of food intake, fertility, blood glucose, and/or body mass. There was evidence of interactions between reproductive traits and hormones related to energy balance and reproduction, but this study did not find evidence that variation in these hormones caused variation in reproductive phenotype.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Leptin from mouse, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder