Merck
  • Mutations in the tricarboxylic acid cycle enzyme, aconitase 2, cause either isolated or syndromic optic neuropathy with encephalopathy and cerebellar atrophy.
Mutations in the tricarboxylic acid cycle enzyme, aconitase 2, cause either isolated or syndromic optic neuropathy with encephalopathy and cerebellar atrophy.
Journal of medical genetics (2014-10-30)
Metodi Dimitrov Metodiev, Sylvie Gerber, Laurence Hubert, Agnès Delahodde, Dominique Chretien, Xavier Gérard, Patrizia Amati-Bonneau, Marie-Christine Giacomotto, Nathalie Boddaert, Anna Kaminska, Isabelle Desguerre, Jeanne Amiel, Marlène Rio, Josseline Kaplan, Arnold Munnich, Agnès Rötig, Jean Michel Rozet, Claude Besmond
RESUMEN
Inherited optic neuropathy has been ascribed to mutations in mitochondrial fusion/fission dynamics genes, nuclear and mitochondrial DNA-encoded respiratory enzyme genes or nuclear genes of poorly known mitochondrial function. However, the disease causing gene remains unknown in many families. We used exome sequencing in order to identify the gene responsible for isolated or syndromic optic atrophy in five patients from three independent families. We found homozygous or compound heterozygous missense and frameshift mutations in the gene encoding mitochondrial aconitase (ACO2), a tricarboxylic acid cycle enzyme, catalysing interconversion of citrate into isocitrate. Unlike wild type ACO2, all mutant ACO2 proteins failed to complement the respiratory growth of a yeast aco1-deletion strain. Retrospective studies using patient-derived cultured skin fibroblasts revealed various degrees of deficiency in ACO2 activity, but also in ACO1 cytosolic activity. Our study shows that autosomal recessive ACO2 mutations can cause either isolated or syndromic optic neuropathy. This observation identifies ACO2 as the second gene responsible for non-syndromic autosomal recessive optic neuropathies and provides evidence for a genetic overlap between isolated and syndromic forms, giving further support to the view that optic atrophy is a hallmark of defective mitochondrial energy supply.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(±)-Potassium citramalate monohydrate, ≥95.0% (NT)