Merck
beta-endorphin stimulates cytokeratin 16 expression and downregulates mu-opiate receptor expression in human epidermis.
The Journal of investigative dermatology (2000-02-26)
M Bigliardi-Qi, P L Bigliardi, A N Eberle, S Büchner, T Rufli
RESUMEN
It has been reported that opioid peptides modulate the differentiation of normal human keratinocytes and that mu-opiate receptors are expressed in human epidermis. The regulation of keratinocyte differentiation is particularly important in psoriasis, and one of the markers for hyperproliferative and differentiating skin diseases is cytokeratin 16. The finding that the endogenous mu-opiate receptor ligand beta-endorphin is increased in serum of patients with psoriasis indicates that the mu-opiate system may play an important role in the pathophysiology of the skin. In this study, we addressed the question whether there is a link between mu-opiate receptor regulation and cytokeratin 16 expression in normal and psoriatic skin. Firstly, we demonstrate that beta-endorphin concentrations between 16 and 1000 nM significantly downregulate mu-opiate receptor expression in epidermis of cultured human skin after 48 h. Secondly, we show that beta-endorphin regulates cytokeratin 16 expression in the epidermis of skin organ cultures exposed to 41-125 nM beta-endorphin for 48 h, leading to elevated cytokeratin 16 production. As expected, the expression of cytokeratin 16 was detected primarily in the suprabasal layer. The same pattern was observed in psoriatic lesional skin, i.e., mu-opiate receptor expression was significantly downregulated and cytokeratin 16 expression upregulated. These results suggest that the mu-opiate receptor system and its ligand beta-endorphin are involved in the pathogenesis of psoriasis, especially in terms of differentiation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
β-Endorphin human, ≥95% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad