Merck
A Chinese hamster cDNA encoding a protein essential for phosphatidylserine synthase I activity.
The Journal of biological chemistry (1991-12-15)
O Kuge, M Nishijima, Y Akamatsu
RESUMEN
A phosphatidylserine-auxotrophic mutant of cultured Chinese hamster ovary (CHO) cells, PSA-3, is defective in phosphatidylserine synthase I activity. Transfection of PSA-3 cells with a cDNA expression library of CHO-K1 (the parent of PSA-3) yielded a phosphatidylserine-prototrophic transformant with normal phosphatidylserine synthase I activity. Using a cDNA segment retrieved from the transformant as a probe, a cDNA clone (pssA) responsible for phosphatidylserine prototrophy was isolated from the original cDNA library by colony filter hybridization. Introduction of the pssA cDNA into PSA-3 cells restored the phosphatidylserine content, and the resultant transformant exhibited 15-fold higher specific phosphatidylserine synthase I activity than CHO-K1 cells. The nucleotide sequence of the pssA cDNA contained a single long open reading frame capable of encoding a protein of 471 amino acid residues with several putative membrane-spanning domains. Our results indicated that the pssA cDNA encodes an integral membrane protein essential for phosphatidylserine synthase I activity.