- An effective microwave-induced iodine-catalyzed method for the synthesis of quinoxalines via condensation of 1,2-diamines with 1,2-dicarbonyl compounds.
An effective microwave-induced iodine-catalyzed method for the synthesis of quinoxalines via condensation of 1,2-diamines with 1,2-dicarbonyl compounds.
Molecules (Basel, Switzerland) (2010-07-27)
Debasish Bandyopadhyay, Sanghamitra Mukherjee, Robert R Rodriguez, Bimal K Banik
PMID20657435
RESUMEN
A microwave-induced iodine-catalyzed simple, rapid and convenient synthesis of different types of quinoxalines via condensation of 1,2-diamines with 1,2-dicarbonyl compounds has been accomplished with an excellent yield.