Merck
Genome-wide association study identifies FCGR2A as a susceptibility locus for Kawasaki disease.
Nature genetics (2011-11-15)
Chiea Chuen Khor, Sonia Davila, Willemijn B Breunis, Yi-Ching Lee, Chisato Shimizu, Victoria J Wright, Rae S M Yeung, Dennis E K Tan, Kar Seng Sim, Jie Jin Wang, Tien Yin Wong, Junxiong Pang, Paul Mitchell, Rolando Cimaz, Nagib Dahdah, Yiu-Fai Cheung, Guo-Ying Huang, Wanling Yang, In-Sook Park, Jong-Keuk Lee, Jer-Yuarn Wu, Michael Levin, Jane C Burns, David Burgner, Taco W Kuijpers, Martin L Hibberd
RESUMEN
Kawasaki disease is a systemic vasculitis of unknown etiology, with clinical observations suggesting a substantial genetic contribution to disease susceptibility. We conducted a genome-wide association study and replication analysis in 2,173 individuals with Kawasaki disease and 9,383 controls from five independent sample collections. Two loci exceeded the formal threshold for genome-wide significance. The first locus is a functional polymorphism in the IgG receptor gene FCGR2A (encoding an H131R substitution) (rs1801274; P = 7.35 × 10(-11), odds ratio (OR) = 1.32), with the A allele (coding for histadine) conferring elevated disease risk. The second locus is at 19q13, (P = 2.51 × 10(-9), OR = 1.42 for the rs2233152 SNP near MIA and RAB4B; P = 1.68 × 10(-12), OR = 1.52 for rs28493229 in ITPKC), which confirms previous findings(1). The involvement of the FCGR2A locus may have implications for understanding immune activation in Kawasaki disease pathogenesis and the mechanism of response to intravenous immunoglobulin, the only proven therapy for this disease.