Merck
  • Surface Acidity and Hydrophilicity of Coprecipitated Al(2)O(3)-SiO(2) Xerogels Prepared from Aluminium Nitrate Nonahydrate and Tetraethylorthosilicate.
Surface Acidity and Hydrophilicity of Coprecipitated Al(2)O(3)-SiO(2) Xerogels Prepared from Aluminium Nitrate Nonahydrate and Tetraethylorthosilicate.
Journal of colloid and interface science (1999-10-21)
Okada, Tomita, Kameshima, Yasumori, MacKenzie
RESUMEN
Amorphous aluminosilicate xerogels with various chemical compositions were prepared by coprecipitation, and their surface acidity and hydrophilicity were investigated by NH(3) gas temperature programed desorption (TPD), water vapor adsorption-desorption isotherms, and (27)Al magic angle spinning nuclear magnetic resonance (MAS NMR). The xerogels were synthesized by adding conc. NH(4)OH to an ethanol solution of calculated amounts of aluminium nitrate nonahydrate and tetraethylorthosilicate, and calcined at 300 degrees C for 4 h. All the NH(3) TPD spectra of the xerogels showed similar asymmetric peak profiles at around 200 degrees C tailing to the higher temperature side. The amount of acidity evaluated from the peak area of the TPD spectra showed a maximum at around 10 mol% Al(2)O(3) composition. The change as a function of composition showed a good correlation with the total amount of four and five coordinated Al atoms in the xerogels deduced from the (27)Al MAS NMR spectra. The water vapor adsorption isotherms of the xerogels were all of type IV irrespective of the composition. The maximum amounts of water vapor adsorbed by these xerogels were about 600-700 ml(STP)/g and were relatively high compared with those for various other adsorbents reported so far. Since the thickness of the adsorbed water vapor layer of the xerogels in the low relative pressure region increased with increasing Al(2)O(3) content, the surface of the xerogels is considered to become more hydrophilic with increasing Al(2)O(3) content of the xerogels. Copyright 1999 Academic Press.