Merck
  • Soluble endoglin specifically binds bone morphogenetic proteins 9 and 10 via its orphan domain, inhibits blood vessel formation, and suppresses tumor growth.
Soluble endoglin specifically binds bone morphogenetic proteins 9 and 10 via its orphan domain, inhibits blood vessel formation, and suppresses tumor growth.
The Journal of biological chemistry (2011-07-09)
Roselyne Castonguay, Eric D Werner, Robert G Matthews, Eleonora Presman, Aaron W Mulivor, Nicolas Solban, Dianne Sako, R Scott Pearsall, Kathryn W Underwood, Jasbir Seehra, Ravindra Kumar, Asya V Grinberg
RESUMEN
Endoglin (CD105), a transmembrane protein of the transforming growth factor β superfamily, plays a crucial role in angiogenesis. Mutations in endoglin result in the vascular defect known as hereditary hemorrhagic telangiectasia (HHT1). The soluble form of endoglin was suggested to contribute to the pathogenesis of preeclampsia. To obtain further insight into its function, we cloned, expressed, purified, and characterized the extracellular domain (ECD) of mouse and human endoglin fused to an immunoglobulin Fc domain. We found that mouse and human endoglin ECD-Fc bound directly, specifically, and with high affinity to bone morphogenetic proteins 9 and 10 (BMP9 and BMP10) in surface plasmon resonance (Biacore) and cell-based assays. We performed a function mapping analysis of the different domains of endoglin by examining their contributions to the selectivity and biological activity of the protein. The BMP9/BMP10 binding site was localized to the orphan domain of human endoglin composed of the amino acid sequence 26-359. We established that endoglin and type II receptors bind to overlapping sites on BMP9. In the in vivo chick chorioallantoic membrane assay, the mouse and the truncated human endoglin ECD-Fc both significantly reduced VEGF-induced vessel formation. Finally, murine endoglin ECD-Fc acted as an anti-angiogenic factor that decreased blood vessel sprouting in VEGF/FGF-induced angiogenesis in in vivo angioreactors and reduced the tumor burden in the colon-26 mouse tumor model. Together our findings indicate an important role of soluble endoglin ECD in the regulation of angiogenesis and highlight efficacy of endoglin-Fc as a potential anti-angiogenesis therapeutic agent.