Merck
  • Mononuclear copper(II) nitrato complexes with methyl-substituted 4-nitropyridine N-oxide. Physicochemical and cytotoxic characteristics.
Mononuclear copper(II) nitrato complexes with methyl-substituted 4-nitropyridine N-oxide. Physicochemical and cytotoxic characteristics.
Journal of inorganic biochemistry (2009-11-26)
Aniela Puszko, Anna Krojcer, Marzena Pełczynska, Joanna Wietrzyk, Maria Cieślak-Golonka, Julia Jezierska, Anna Adach, Maria Kubiak
RESUMEN
Three new complexes, products of the interaction of Cu(NO(3))(2) and methyl-substituted 4-nitropyridine N-oxides were synthesized and characterized by elemental analysis, magnetic, spectroscopic (IR, FIR and EPR), thermal and X-ray methods. The complexes (magnetic moments 1.70-1.81 BM at 300K) of general formula [Cu(H(2)O)(NO(3))(2)L(2)], L=2-methyl-4-nitropyridine N-oxide and [Cu(NO(3))(2) L'(2)], where L'=2,6-dimethyl- and 2,3,6-trimethyl-4-nitropyridine N-oxide were obtained. The compounds were unstable upon dissolution. The X-ray single crystal structure of Cu(II) complex with 2,6-dimethyl-4-nitropyridine N-oxide was determined and analysed. The compounds and free ligands were tested in vitro on the cytotoxic activity against MCF-7 and SW-707 human cancer cell lines. The complexes with 4-nitropyridine N-oxide (a reference) and 2-methyl-4-nitropyridine N-oxide show a significant anti-proliferative activity against studied cell lines. A reciprocal relationship between the activity and the number of methyl groups was observed. Both ligands and complexes are cytotoxic active but to the different cell lines.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Methyl-4-nitropyridine N-oxide, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad