Merck
Single-walled carbon nanohorns coated with Fe2O3 as a superior anode material for lithium ion batteries.
Chemical communications (Cambridge, England) (2011-06-01)
Yi Zhao, Jiaxin Li, Yunhai Ding, Lunhui Guan
RESUMEN
A novel composite of Fe(2)O(3) and single-walled carbon nanohorns (SWCNHs) was firstly developed via a simple hydrothermal method. As an anode material for lithium ion batteries, a Fe(2)O(3)/SWCNHs composite shows excellent rate performance and cycle stability, even at a high current density of 1000 mA g(-1).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Carbon nanohorns, as-grown
Sigma-Aldrich
Carbon nanohorns, oxidized, avg. no. of layers, 1