Merck
  • The bromodomain protein Brd4 is a positive regulatory component of P-TEFb and stimulates RNA polymerase II-dependent transcription.
The bromodomain protein Brd4 is a positive regulatory component of P-TEFb and stimulates RNA polymerase II-dependent transcription.
Molecular cell (2005-08-20)
Moon Kyoo Jang, Kazuki Mochizuki, Meisheng Zhou, Ho-Sang Jeong, John N Brady, Keiko Ozato
RESUMEN
Brd4 is a mammalian bromodomain protein that binds to acetylated chromatin. Proteomic analysis revealed that Brd4 interacts with cyclinT1 and Cdk9 that constitutes core positive transcription elongation factor b (P-TEFb). Brd4 interacted with P-TEFb in the living nucleus through its bromodomain. About half of P-TEFb was bound to the inhibitory subunit and functionally inactive. Brd4 interacted with P-TEFb that was free of the inhibitory subunit. An increase in Brd4 expression led to increased P-TEFb-dependent phosphorylation of RNA polymerase II (RNAPII) CTD and stimulation of transcription from promoters in vivo. Conversely, a reduction in Brd4 expression by siRNA reduced CTD phosphorylation and transcription, revealing that Brd4 is a positive regulatory component of P-TEFb. In chromatin immunoprecipitation (ChIP) assays, the recruitment of P-TEFb to a promoter was dependent on Brd4 and was enhanced by an increase in chromatin acetylation. Together, P-TEFb alternately interacts with Brd4 and the inhibitory subunit to maintain functional equilibrium in the cell.