Merck
  • Five SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin (SMARC) genes are dispersed in the human genome.
Five SWI/SNF-related, matrix-associated, actin-dependent regulator of chromatin (SMARC) genes are dispersed in the human genome.
Genomics (1998-08-07)
H Z Ring, V Vameghi-Meyers, W Wang, G R Crabtree, U Francke
RESUMEN
The SWI/SNF-related, matrix-associated, actin-dependent regulators of chromatin (SMARC), also called BRG1-associated factors, are components of human SWI/SNF-like chromatin-remodeling protein complexes. We mapped five human SMARC genes toregions on four different human chromosomes, SMARCC1 to 3p23-p21, SMARCC2 to 12q13-q14, SMARCD1 to 12q13-q14, SMARCD2 to 17q23-q24, and SMARCD3 to 7q35-q36. SMARCC1, SMARCC2, and SMARCD1 are assigned to chromosomal regions that are frequently involved in somatic rearrangements in human cancers. SMARCD1 was mapped to the critical region of Allgrove syndrome; however, no mutation was identified in one Allgrove syndrome family studied.