Merck
Transcription activating property of autoantigen SG2NA and modulating effect of WD-40 repeats.
Experimental cell research (2001-09-26)
W Zhu, E K Chan, J Li, P Hemmerich, E M Tan
RESUMEN
Autoantibodies to intracellular proteins have been detected in sera of patients with various forms of cancer. Nuclear autoantigen SG2NA (S, G2 phase nuclear antigen) was isolated using autoantibodies from a patient with bladder and lung cancers and its expression is enhanced in the S and G2 phases of the cell cycle. Molecular cloning revealed that the C-terminal region of SG2NA contains six WD-40 repeats, motifs that are present in a large family of proteins with diverse functions. We show that the N-terminal region of SG2NA (aa 1-391) acted as a strong transcriptional activator in both yeast and mammalian cells. In contrast, the C-terminal WD-40 repeats had an inhibitory effect on transcription activation. We performed molecular swapping experiments by substituting the WD-40 repeats of SG2NA with those of yeast Met30 and Cdc4 and showed that the WD-40 regions from either Met30 or Cdc4 were capable of reproducing transcription repression function. The SG2NA WD-40 repeats were also able to repress basal level transcription and transactivation function of a GAL4-VP16 chimera. These observations suggest that some WD-40 repeats may have, as one of their functions, a negative regulatory role in the biological activities of their own and perhaps other proteins.