Merck
Overexpression of high-mobility group box 2 is associated with tumor aggressiveness and prognosis of hepatocellular carcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-09-21)
Jung-Hee Kwon, Jongmin Kim, Jin Young Park, Sun Mi Hong, Chang Wook Park, Seok Joo Hong, Sun Young Park, Yoon Jung Choi, In-Gu Do, Jae-Won Joh, Dae Shick Kim, Kwan Yong Choi
RESUMEN
We investigated the expression of high-mobility group box 2 (HMGB2) in patients with hepatocellular carcinoma (HCC) and its clinical effects with underlying mechanisms. HMGB2 mRNA levels were measured in 334 HCC patients by real-time reverse transcription-PCR and HMGB2 protein levels in 173 HCC patients by immunohistochemical studies. The HMGB2 expression level was measured by Western blotting for three HCC cell lines. To clarify the precise role of HMGB2 on cell proliferation, we did in vitro analysis with expression vectors and small interfering RNAs. HMGB2 mRNA and protein expression were significantly higher in HCC than in noncancerous surrounding tissues (P < 0.0001) and showed a positive correlation (ρ = 0.35, P < 0.001). HMGB2 overexpression was significantly correlated with shorter overall survival time, both at mRNA (P = 0.0054) and protein level (P = 0.023). Moreover, HMGB2 mRNA level was an independent prognostic factor for overall survival in a multivariate analysis (P = 0.0037). HMGB2 knockdown by small interfering RNAs decreased cell proliferation, and overexpression of HMGB2 by expression vectors diminished cisplatin- and etoposide-induced cell death. Our clinical and in vitro data suggest that HMGB2 plays a significant role in tumor development and prognosis of HCC. These results can partly be explained by altered cell proliferations by HMGB2 associated with the antiapoptotic pathway.