Merck
Allele-selective inhibition of mutant atrophin-1 expression by duplex and single-stranded RNAs.
Biochemistry (2014-07-02)
Jiaxin Hu, Jing Liu, K Jayaprakash Narayanannair, Jeremy G Lackey, Satya Kuchimanchi, Kallanthottathil G Rajeev, Muthiah Manoharan, Eric E Swayze, Walt F Lima, Thazha P Prakash, Qin Xiang, Carlos Martinez, David R Corey
RESUMEN
Dentatorubral-pallidoluysian atrophy (DRPLA) is a progressive neurodegenerative disorder that currently has no curative treatments. DRPLA is caused by an expansion of a CAG trinucleotide repeat region within the protein-encoding sequence of the atrophin-1 (ATN-1) gene. Inhibition of mutant ATN-1 protein expression is one strategy for treating DRPLA, and allele-selective gene silencing agents that block mutant expression over wild-type expression would be lead compounds for therapeutic development. Here we develop an assay for distinguishing mutant from wild-type ATN-1 protein by gel electrophoresis. We use this assay to evaluate duplex RNAs and single-stranded silencing RNAs (ss-siRNAs) for allele-selective inhibition of ATN-1 protein expression. We observed potent and allele-selective inhibition by RNA duplexes that contain mismatched bases relative to the CAG target and have the potential to form miRNA-like complexes. ss-siRNAs that contained mismatches were as selective as mismatch-containing duplexes. We also report allele-selective inhibition by duplex RNAs containing unlocked nucleic acids or abasic substitutions, although selectivities are not as high. Five compounds that showed >8-fold allele selectivity for mutant ATN-1 were also selective for inhibiting the expression of two other trinucleotide repeat disease genes, ataxin-3 (ATXN-3) and huntingtin (HTT). These data demonstrate that the expanded trinucleotide repeat within ATN-1 mRNA is a potential target for compounds designed to achieve allele-selective inhibition of ATN-1 protein, and one agent may allow the targeting of multiple disease genes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Medio esencial mínimo Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture