Merck
Analysis of lignocellulose derived phenolic monomers by headspace solid-phase microextraction and gas chromatography.
Journal of chromatography. A (2013-08-13)
Michaela Kolb, Doris Schieder, Martin Faulstich, Volker Sieber
RESUMEN
A headspace solid-phase microextraction method with subsequent GC-MS (HS-SPME/GC-MS) was established for the quantitative analysis of volatile lignin derived phenolic monomers in complex aqueous solutions. Extraction was done using a polyacrylate fiber. The optimization of HS-SPME - parameters was performed using a multi component model solution of six representative phenolic monomers identified in liquid hot water (LHW) supernatants of hydrothermally treated lignocellulosic biomass: p-coumaric acid, guaiacol, vanillin, acetosyringone, 4-hydroxy-3-methoxyphenylacetone, and acetophenone. Plackett-Burman design was applied for pre-evaluation and 2(3) central composite designs with star points for parameter optimization. LOQ (S/N>10) and LOD (S/N>3) were determined for 12 phenols yielding LOQ of <0.005-618nM and LOD of <0.005-412nM. Within-day and between-day tests (n=6) showed different results for the tested phenols. RSD ranged from 2% to 30% and recovery rates from 99% to 160% in LHW matrix. Tests on storage of LHW supernatants for several weeks indicated a considerable influence of temperature on the stability of the solutions which may even have to be taken into account for auto sampler handling. All in all the method allows a fast and solvent free analysis requiring low sample volumes making it a powerful tool for screening or high-throughput analysis of aqueous solutions of lignin derived aromatics.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Hydroxy-3-methoxyphenylacetone, 96%