Merck
The requirement of the DEAD-box protein DDX24 for the packaging of human immunodeficiency virus type 1 RNA.
Virology (2008-02-22)
Jing Ma, Liwei Rong, Yongdong Zhou, Bibhuti Bushan Roy, Jennifer Lu, Levon Abrahamyan, Andrew J Mouland, Qinghua Pan, Chen Liang
RESUMEN
RNA helicases play important roles in RNA metabolism. Human immunodeficiency virus type 1 (HIV-1) does not carry its own RNA helicase, the virus thus needs to exploit cellular RNA helicases to promote the replication of its RNA at various steps such as transcription, folding and transport. In this study, we report that knockdown of a DEAD-box protein named DDX24 inhibits the packaging of HIV-1 RNA and thus diminishes viral infectivity. The decreased viral RNA packaging as a result of DDX24-knockdown is observed only in the context of the Rev/RRE (Rev response element)-dependent but not the CTE (constitutive transport element)-mediated nuclear export of viral RNA, which is explained by the specific interaction of DDX24 with the Rev protein. We propose that DDX24 acts at the early phase of HIV-1 RNA metabolism prior to nuclear export and the consequence of this action extends to the viral RNA packaging stage during virus assembly.