Merck
A chaperone-like function of intramolecular high-mannose chains in the oxidative refolding of bovine pancreatic RNase B.
Journal of biochemistry (1996-09-01)
H Yamaguchi, M Uchida
RESUMEN
This paper describes a chaperone-like function of the intramolecular N-glycans of bovine pancreatic RNase B. We studied air-oxidative regeneration from reductively denatured species of RNase B and its nonglycosylated form, RNase A. RNase B was reactivated much faster than RNase A, while RNase A was liable to aggregate during the regeneration. An Asn-linked oligosaccharide, Man5GlcNA2Asn, which corresponds to the most predominant sugar chain (ca. 60%) of RNase B, enhanced the reactivation of the denatured RNases A and B. The stimulatory effect of this Asn-oligosaccharide revealed that the N-glycans of RNase B facilitate the transformation of bulky intermediates into folded, compact species.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)