Merck
1-Methyl-1H-benzimidazole-2(3H)-thione.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Hizbullah Khan, Amin Badshah, Farkhanda Shaheen, Christine Gieck, Rizwana Aleem Qureshi
RESUMEN
The title compound, C(8)H(8)N(2)S, was prepared by the condensation of N-methyl-1,2-phenyl-enediamine and carbon disulfide. The crystal structure is stabilized by a C-H⋯π inter-action between a benzene H atom and the benzene ring of a neighbouring mol-ecule, and by inter-molecular N-H⋯S inter-actions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N-Methyl-1,2-phenylenediamine, 97%