Merck
  • Clinical value of circulating endothelial cells and of soluble CD146 levels in patients undergoing surgery for non-small cell lung cancer.
Clinical value of circulating endothelial cells and of soluble CD146 levels in patients undergoing surgery for non-small cell lung cancer.
British journal of cancer (2014-01-30)
M Ilie, E Long, V Hofman, E Selva, C Bonnetaud, J Boyer, N Vénissac, C Sanfiorenzo, B Ferrua, C-H Marquette, J Mouroux, P Hofman
RESUMEN
Previous studies indicate that endothelial injury, as demonstrated by the presence of circulating endothelial cells (CECs), may predict clinical outcome in cancer patients. In addition, soluble CD146 (sCD146) may reflect activation of angiogenesis. However, no study has investigated their combined clinical value in patients undergoing resection for non-small cell lung cancer (NSCLC). Data were collected from preoperative blood samples from 74 patients who underwent resection for NSCLC. Circulating endothelial cells were defined, using the CellSearch Assay, as CD146+CD105+CD45-DAPI+. In parallel, sCD146 was quantified using an ELISA immunoassay. These experiments were also performed on a group of 20 patients with small-cell lung cancer, 60 healthy individuals and 23 patients with chronic obstructive pulmonary disease. The CEC count and the plasma level of sCD146 were significantly higher in NSCLC patients than in the sub-groups of controls (P<0.001). Moreover, an increased CEC count was associated with higher levels of sCD146 (P=0.010). Both high CEC count and high sCD146 plasma level at baseline significantly correlated with shorter progression-free survival (P<0.001, respectively) and overall survival (P=0.005; P=0.009) of NSCLC patients. The present study provides supportive evidence to show that both a high CEC count and a high sCD146 level at baseline correlate with poor prognosis and may be useful for the prediction of clinical outcome in patients undergoing surgery for NSCLC.