Merck
Inhibition of hepatitis C virus replication by a specific inhibitor of serine-arginine-rich protein kinase.
Antimicrobial agents and chemotherapy (2010-05-26)
Yuko Karakama, Naoya Sakamoto, Yasuhiro Itsui, Mina Nakagawa, Megumi Tasaka-Fujita, Yuki Nishimura-Sakurai, Sei Kakinuma, Masaya Oooka, Seishin Azuma, Kiichiro Tsuchiya, Hiroshi Onogi, Masatoshi Hagiwara, Mamoru Watanabe
RESUMEN
Splicing of messenger RNAs is regulated by site-specific binding of members of the serine-arginine-rich (SR) protein family, and SR protein kinases (SRPK) 1 and 2 regulate overall activity of the SR proteins by phosphorylation of their RS domains. We have reported that specifically designed SRPK inhibitors suppressed effectively several DNA and RNA viruses in vitro and in vivo. Here, we show that an SRPK inhibitor, SRPIN340, suppressed in a dose-dependent fashion expression of a hepatitis C virus (HCV) subgenomic replicon and replication of the HCV-JFH1 clone in vitro. The inhibitory effects were not associated with antiproliferative or nonspecific cytotoxic effects on the host cells. Overexpression of SRPK1 or SRPK2 resulted in augmentation of HCV replication, while small interfering RNA (siRNA) knockdown of the SRPKs suppressed HCV replication significantly. Immunocytochemistry showed that SRPKs and the HCV core and NS5A proteins colocalized to some extent in the perinuclear area. Our results demonstrate that SRPKs are host factors essential for HCV replication and that functional inhibitors of these kinases may constitute a new class of antiviral agents against HCV infection.