Merck
  • Synthesis of 5α-androstane-17-spiro-δ-lactones with a 3-keto, 3-hydroxy, 3-spirocarbamate or 3-spiromorpholinone as inhibitors of 17β-hydroxysteroid dehydrogenases.
Synthesis of 5α-androstane-17-spiro-δ-lactones with a 3-keto, 3-hydroxy, 3-spirocarbamate or 3-spiromorpholinone as inhibitors of 17β-hydroxysteroid dehydrogenases.
Molecules (Basel, Switzerland) (2013-01-25)
Guy Bertrand Djigoué, Béatrice Tchédam Ngatcha, Jenny Roy, Donald Poirier
RESUMEN
We synthesized two series of androstane derivatives as inhibitors of type 3 and type 5 17β-hydroxysteroid dehydrogenases (17β-HSDs). In the first series, four monospiro derivatives at position C17 were prepared from androsterone (ADT) or epi-ADT. After the protection of the alcohol at C3, the C17-ketone was alkylated with the lithium acetylide of tetrahydro-2-(but-3-ynyl)-2-H-pyran, the triple bond was hydrogenated, the protecting groups hydrolysed and the alcohols oxidized to give the corresponding 3-keto-17-spiro-lactone derivative. The other three compounds were generated from this keto-lactone by reducing the ketone at C3, or by introducing one or two methyl groups. In the second series, two dispiro derivatives at C3 and C17 were prepared from epi-ADT. After introducing a spiro-δ-lactone at C17 and an oxirane at C3, an aminolysis of the oxirane with L-isoleucine methyl ester provided an amino alcohol, which was treated with triphosgene or sodium methylate to afford a carbamate- or a morpholinone-androstane derivative, respectively. These steroid derivatives inhibited 17β-HSD3 (14-88% at 1 μM; 46-94% at 10 μM) and 17β-HSD5 (54-73% at 0.3 μM; 91-92% at 3 μM). They did not produce any androgenic activity and did not bind steroid (androgen, estrogen, glucocorticoid and progestin) receptors, suggesting a good profile for prostate cancer therapy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-(3-Butynyloxy)tetrahydro-2H-pyran, 97%