Merck
Asymmetric reduction of a ketone by wet and lyophilized cells of Geotrichum candidum in organic solvents.
New biotechnology (2011-08-13)
Mani Shankar Bhattacharyya, Amit Singh, Uttam Chand Banerjee
RESUMEN
Sixteen organic co-solvents were screened for stereoselective reduction of 1-acetonapthone in aqueous media by whole cells of Geotrichum candidum. Benzyl alcohol was found to be a good co-solvent as it afforded a high coversion and reduced deactivation of the cells. Half-lives of the wet and lyophilized whole cell biocatalysts in pure benzyl alcohol were 23.07 and 11.21 hours, respectively. The initial reaction rates at 30°C were 13.1 and 11.0μmol/min, respectively, for the wet and lyophilized cells. With optimized conditions in a reaction medium containing phosphate buffer and benzyl alcohol (1:1 by vol) with 230mM 1-acetonapthone, more than 98% and 81% conversion (ee >99%) was achieved in 5 hours with the wet and lyophilized cells, respectively. Both the cell preparations showed maximum conversion at 30°C. A thermodynamic characterization revealed that the wet cells were more thermostable than the lyophilized cells. The calculated half-life of the wet cells at pH 7 was 93 hours, whereas that of the lyophilized cells was 71 hours at the same condition.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Acetonaphthone, 97%