- Nonpeptide glycoprotein IIb/IIIa inhibitors: substituted quinazolinediones and quinazolinones as potent fibrinogen receptor antagonists.
Nonpeptide glycoprotein IIb/IIIa inhibitors: substituted quinazolinediones and quinazolinones as potent fibrinogen receptor antagonists.
Bioorganic & medicinal chemistry letters (1999-01-01)
N J Liverton, D J Armstrong, D A Claremon, D C Remy, J J Baldwin, R J Lynch, G Zhang, R J Gould
PMID9871603
RESUMEN
The synthesis and biological activity of a series of 3,6-substituted quinazolinediones and quinazolinones are described. The potent activity of these compounds as platelet aggregation inhibitors demonstrates the utility of these structures as central templates for nonpeptide RGD mimics.