Merck
Antidiabetic actions of an estrogen receptor β selective agonist.
Diabetes (2013-01-26)
Paloma Alonso-Magdalena, Ana B Ropero, Marta García-Arévalo, Sergi Soriano, Iván Quesada, Sarheed J Muhammed, Albert Salehi, Jan-Ake Gustafsson, Angel Nadal
RESUMEN
The estrogen receptor β (ERβ) is emerging as an important player in the physiology of the endocrine pancreas. We evaluated the role and antidiabetic actions of the ERβ selective agonist WAY200070 as an insulinotropic molecule. We demonstrate that WAY200070 enhances glucose-stimulated insulin secretion both in mouse and human islets. In vivo experiments showed that a single administration of WAY200070 leads to an increase in plasma insulin levels with a concomitant improved response to a glucose load. Two-week treatment administration increased glucose-induced insulin release and pancreatic β-cell mass and improved glucose and insulin sensitivity. In addition, streptozotocin-nicotinamide-induced diabetic mice treated with WAY200070 exhibited a significant improvement in plasma insulin levels and glucose tolerance as well as a regeneration of pancreatic β-cell mass. Studies performed in db/db mice demonstrated that this compound restored first-phase insulin secretion and enhanced pancreatic β-cell mass. We conclude that ERβ agonists should be considered as new targets for the treatment of diabetes.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
WAY-200070, ≥98% (HPLC)