Merck
Pathways for Neoarchean pyrite formation constrained by mass-independent sulfur isotopes.
Proceedings of the National Academy of Sciences of the United States of America (2013-02-15)
James Farquhar, John Cliff, Aubrey L Zerkle, Alexey Kamyshny, Simon W Poulton, Mark Claire, David Adams, Brian Harms
RESUMEN
It is generally thought that the sulfate reduction metabolism is ancient and would have been established well before the Neoarchean. It is puzzling, therefore, that the sulfur isotope record of the Neoarchean is characterized by a signal of atmospheric mass-independent chemistry rather than a strong overprint by sulfate reducers. Here, we present a study of the four sulfur isotopes obtained using secondary ion MS that seeks to reconcile a number of features seen in the Neoarchean sulfur isotope record. We suggest that Neoarchean ocean basins had two coexisting, significantly sized sulfur pools and that the pathways forming pyrite precursors played an important role in establishing how the isotopic characteristics of each of these pools was transferred to the sedimentary rock record. One of these pools is suggested to be a soluble (sulfate) pool, and the other pool (atmospherically derived elemental sulfur) is suggested to be largely insoluble and unreactive until it reacts with hydrogen sulfide. We suggest that the relative contributions of these pools to the formation of pyrite depend on both the accumulation of the insoluble pool and the rate of sulfide production in the pyrite-forming environments. We also suggest that the existence of a significant nonsulfate pool of reactive sulfur has masked isotopic evidence for the widespread activity of sulfate reducers in the rock record.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Iron disulfide, powder, −325 mesh, 99.8% trace metals basis