- Synthesis and structure-immunosuppressive activity relationships of bakuchiol and its derivatives.
Synthesis and structure-immunosuppressive activity relationships of bakuchiol and its derivatives.
Bioorganic & medicinal chemistry (2007-12-08)
Hongli Chen, Xiaolong Du, Wei Tang, Yu Zhou, Jianping Zuo, Huijin Feng, Yuanchao Li
PMID18063371
RESUMEN
A series of derivatives of bakuchiol were synthesized and tested in vitro for their cytotoxicity, and inhibition of T cell proliferation and B cell proliferation. The data obtained provided preliminary structure-activity relationships of the compounds as immunosuppressive activity.