Merck
Protective effect of calceolarioside on adriamycin-induced cardiomyocyte toxicity.
European journal of pharmacology (2006-06-20)
Do-Sung Kim, Hyung-Ryong Kim, Eun-Rhan Woo, Dae-Young Kwon, Myung-Sunny Kim, Soo-Wan Chae, Han-Jung Chae
RESUMEN
Adriamycin is a potent antitumor drug that is known to cause severe cardiotoxicity. This study examined the protective effect of calceolarioside on adriamycin-induced cardiomyocyte toxicity. Calceolarioside significantly inhibited the adriamycin induced cell death and caspase-3 activation, which may be explained by the increase in Bcl-2 expression and the inhibition of Bax expression. Calceolarioside increased the expression of the antioxidant molecules and decreased the level of intracellular reactive oxygen species. Catalase, glutathione, N-acetylcysteine, Mannitol and Mn-TBAP (manganese (III) tetrakis-(4-benzoic acid) porphyrin) significantly inhibited the H9c2 cell death induced by adriamycin. Calceolarioside significantly inhibited H9c2 cell death, and was more effective than that observed with the other antioxidants, including probucol, ascorbic acid, and alpha-tocopherol. Overall, these results suggest that calceolarioside can inhibit adriamycin-induced apoptosis in H9c2 cardiomyocyte by inhibiting the generation of reactive oxygen species. Calceolarioside may be a potential candidate agent that inhibits cardiomyocyte-toxicity in adriamycin-exposed patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Calceolarioside A, ≥95% (LC/MS-ELSD)