Merck
  • Differentiation in a rat PC12 cell line induced by ostruthin and (-)-bornyl ferulate, constituents of a Chinese herbal medicine.
Differentiation in a rat PC12 cell line induced by ostruthin and (-)-bornyl ferulate, constituents of a Chinese herbal medicine.
Bioscience, biotechnology, and biochemistry (1999-09-29)
J Qi, M Ojika, Y Sakagami
RESUMEN
A search for neuritogenic compounds in Chinese herbs resulted in the isolation of two known substances, ostruthin and (-)-bornyl ferulate, from Notopterygium incisum (and/or N. forbesii). Both compounds induced comparable neurite-like structures in 20% of rat PC12 cells at 2 micrograms/ml, and showed cytotoxicity at concentrations higher than 3 micrograms/ml.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
6-Geranyl-7-hydroxycoumarin, ≥95% (LC/MS-ELSD)