Merck
hPuf-A/KIAA0020 modulates PARP-1 cleavage upon genotoxic stress.
Cancer research (2011-01-27)
Hao-Yen Chang, Chi-Chen Fan, Po-Chen Chu, Bo-En Hong, Hyeon Jeong Lee, Mau-Sun Chang
RESUMEN
Human hPuf-A/KIAA0020 was first identified as a new minor histocompatibility antigen in 2001. Its zebrafish orthologue contains six Pumilio-homology RNA-binding domains and has been shown to participate in the development of eyes and primordial germ cells, but the cellular function of hPuf-A remains unclear. In this report, we showed that hPuf-A predominantly localized in the nucleoli with minor punctate signals in the nucleoplasm. The nucleolar localization of hPuf-A would redistribute to the nucleoplasm after the treatment of RNA polymerase inhibitors (actinomycin D and 5,6-dichlorobenzimidazole riboside) and topoisomerase inhibitors [camptothecin (CPT) and etoposide]. Interestingly, knockdown of hPuf-A sensitized cells to CPT and UV treatment and cells constitutively overexpressing hPuf-A became more resistant to genotoxic exposure. Affinity gel pull-down coupled with mass spectrometric analysis identified PARP-1 as one of the hPuf-A interacting proteins. hPuf-A specifically interacts with the catalytic domain of PARP-1 and inhibits poly(ADP-ribosyl)ation of PARP-1 in vitro. Depletion of hPuf-A increased the cleaved PARP-1 and overexpression of hPuf-A lessened PARP-1 cleavage when cells were exposed to CPT and UV light. Collectively, hPuf-A may regulate cellular response to genotoxic stress by inhibiting PARP-1 activity and thus preventing PARP-1 degradation by caspase-3.