Merck
Highly active copper-network catalyst for the direct aldol reaction.
Chemistry, an Asian journal (2011-07-14)
Hidetoshi Ohta, Yasuhiro Uozumi, Yoichi M A Yamada
RESUMEN
The development of a highly active solid-phase catechol-copper network catalyst for direct aldol reaction is described. The catalyst was prepared from an alkyl-chain-linked bis(catechol) and a copper(II) complex. The direct aldol reaction between carbonyl compounds (aldehydes and ketones) and methyl isocyanoacetate was carried out using 0.1-1 mol% [Cu] catalyst to give the corresponding oxazolines at yields of up to 99% and a trans/cis ratio of >99:1. The catalyst was reused with no loss of catalytic activity. A plausible reaction pathway is also described.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methyl isocyanoacetate, technical grade, 95%