Merck
  • Electrochemical preparation of tris(tert-butyldimethylsilyl)cyclopropene and its hydride abstraction to tris(tert-butyldimethylsilyl)cyclopropenium tetrafluoroborate.
Electrochemical preparation of tris(tert-butyldimethylsilyl)cyclopropene and its hydride abstraction to tris(tert-butyldimethylsilyl)cyclopropenium tetrafluoroborate.
Proceedings of the National Academy of Sciences of the United States of America (1999-09-01)
H A Buchholz, G K Prakash, D Deffieux, G A Olah
RESUMEN
Electrochemical reductive tert-butyldimethylsilylation of tetrachlorocyclopropene to 1,2,3-tris(tert-butyldimethylsilyl)cyclopropene, a potential strained precursor for Diels-Alder and related cycloaddition reactions, is described. By hydride abstraction with nitrosonium tetrafluoroborate, 1,2,3-tris(tert-butyldimethylsilyl)cyclopropene is ionized quantitatively to Hückeloid 2pi aromatic tris(tert-butyldimethylsilyl)cyclopropenium tetrafluoroborate.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tetrachlorocyclopropene, 98%