Merck
A novel 1,3,5-tris(alpha,beta,beta-trifluorovinyl)benzene monomer.
Chemical communications (Cambridge, England) (2003-11-05)
Lawrence A Ford, Darryl D DesMarteau
RESUMEN
The reaction of the perfluoroalkenylzinc reagent, CF2=CFZnBr, with 1,3,5-tribromobenzene in the presence of a catalytic amount of Pd(Ph3)4 yielded a novel trifunctional monomer 1,3,5-tris(alpha,beta,beta-trifluorovinyl)benzene (1).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,3,5-Tribromobenzene, 98%