Merck
Mononuclear cells enhance prostaglandin E2 production of polymorphonuclear leukocytes via tumor necrosis factor alpha.
Biochemical and biophysical research communications (1990-04-30)
H Akama, Y Ichikawa, Y Matsushita, T Shinozawa, M Homma
RESUMEN
To clarify the interactions between mononuclear cells and polymorphonuclear leukocytes, and to identify the cytokine(s) that mediate the interaction, the effects of a culture supernatant of LPS-stimulated mononuclear cells on production of arachidonic acid metabolites of polymorphonuclear cells were studied. The culture supernatant of LPS-stimulated mononuclear cells increased production of prostaglandin E2 of polymorphonuclear cells. TNF alpha, but not IL-1, IL-2, IL-6, or IFN gamma, enhanced the prostaglandin E2 production when added in vitro. Additionally, an anti-rTNF alpha monoclonal antibody inhibited the stimulating activity of the culture supernatants. TNF alpha, produced by mononuclear cells, appears to play an important role in the development of inflammation, such as rheumatoid arthritis, by enhancing the arachidonic acid metabolism of the polymorphonuclear cells.