Merck
The role of lateral gene transfer in the evolution of isoprenoid biosynthesis pathways.
Molecular microbiology (2000-09-06)
Y Boucher, W F Doolittle
RESUMEN
Lateral gene transfer (LGT) is a major force in microbial genome evolution. Here, we present an overview of lateral transfers affecting genes involved in isopentenyl diphosphate (IPP) synthesis. Two alternative metabolic pathways can synthesize this universal precursor of isoprenoids, the 1-deoxy-D-xylulose 5-phosphate (DOXP) pathway and the mevalonate (MVA) pathway. We have surveyed recent genomic data and the biochemical literature to determine the distribution of the genes composing these pathways within the bacterial domain. The scattered distribution observed is incompatible with a simple scheme of vertical transmission. LGT (among and between bacteria, archaea and eukaryotes) more parsimoniously explains many features of this pattern. This alternative scenario is supported by phylogenetic analyses, which unambiguously confirm several cases of lateral transfer. Available biochemical data allow the formulation of hypotheses about selective pressures favouring transfer. The phylogenetic diversity of the organisms involved and the range of possible causes and effects of these transfer events make the IPP biosynthetic pathways an ideal system for studying the evolutionary role of LGT.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Deoxy-D-xylulose-5-phosphate sodium salt, ≥99.0% (TLC)