- ST segment elevation induced by class IC antiarrhythmic agents: underlying electrophysiologic mechanisms and insights into drug-induced proarrhythmia.
ST segment elevation induced by class IC antiarrhythmic agents: underlying electrophysiologic mechanisms and insights into drug-induced proarrhythmia.
Journal of cardiovascular electrophysiology (1998-12-03)
S C Krishnan, M E Josephson
PMID9835260
RESUMEN
Three patients in whom Class IC sodium channel blockers induced ST segment elevation in leads V1 through V3 are described. The underlying electrophysiologic mechanism, implications for drug-induced proarrhythmia, and the relationship of the finding to the Brugada syndrome type of idiopathic ventricular fibrillation are discussed.