Merck
Retinoid-induced chromatin structure alterations in the retinoic acid receptor beta2 promoter.
Molecular and cellular biology (1997-10-29)
N Bhattacharyya, A Dey, S Minucci, A Zimmer, S John, G Hager, K Ozato
RESUMEN
Transcription of the retinoic acid receptor beta2 (RARbeta2) gene is induced by retinoic acid (RA) in mouse P19 embryonal carcinoma (EC) cells. Here we studied RA-induced chromatin structure alterations in the endogenous RARbeta2 promoter and in an integrated, multicopy RARbeta2 promoter in EC cells. RA markedly increased restriction site accessibility within the promoter, including a site near the RA responsive element (RARE) to which the nuclear receptor retinoid X receptor (RXR)-RAR heterodimer binds. These changes coincided with RA-induced alterations in the DNase I hypersensitivity pattern in and around the promoter. These changes became undetectable upon removal of RA, which coincided with the extinction of transcription. Analyses with receptor-selective ligands and an antagonist showed that increase in restriction site accessibility correlates with transcriptional activation, which parallels the RA-induced in vivo footprint of the promoter. Despite these changes, the micrococcal nuclease digestion profile of this promoter was not altered by RA. These results indicate that concurrent with the binding of the RXR-RAR heterodimer to the RARE, the local chromatin structure undergoes dynamic, reversible changes in and around the promoter without globally affecting the nucleosomal organization.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ro 41-5253, ≥98% (HPLC)