Merck
  • Synthesis and structure-activity relationships of 6,7-benzomorphan derivatives as antagonists of the NMDA receptor-channel complex.
Synthesis and structure-activity relationships of 6,7-benzomorphan derivatives as antagonists of the NMDA receptor-channel complex.
Journal of medicinal chemistry (1997-08-29)
M Grauert, W D Bechtel, H A Ensinger, H Merz, A J Carter
RESUMEN
We have synthesized a series of stereoisomeric 6,7-benzomorphan derivatives with modified N-substituents and determined their ability to antagonize the N-methyl-D-aspartate (NMDA) receptor-channel complex in vitro and in vivo. The ability of the compounds to displace [3H]-MK-801 from the channel site of the NMDA receptor in rat brain synaptosomal membranes and to inhibit NMDA-induced lethality in mice was compared with their ability to bind to the mu opioid receptor. Examination of structure-activity relationships showed that the absolute stereochemistry is critically important for differentiating these two effects. (-)-1R,9 beta,2"S-enantiomers exhibited a higher affinity for the NMDA receptor-channel complex than for the mu opioid receptor. The aromatic hydroxy function was also found to influence the specificity of the compounds. Shift of the hydroxy group from the 2'-position to the 3'-position significantly increased the affinity for the NMDA receptor-channel complex and considerably reduced the affinity for the mu opioid receptor. From this series of 6,7-benzomorphan derivatives, the compound 15cr.HCl [(2R)-[2 alpha, 3(R*),6 alpha]-1,2,3,4,5,6-hexahydro-3-(2-methoxypropyl)-6,11,11-trimethyl -2,6-methano-3-benzazocin-9-ol hydrochloride] was chosen as the optimum candidate for further pharmacological investigations.