Merck
  • Synthesis and evaluation of gastroprotective activity of omeprazole related benzimidazole, imidazo[4,5-b]pyridine and quinoxaline 2-substituted derivatives.
Synthesis and evaluation of gastroprotective activity of omeprazole related benzimidazole, imidazo[4,5-b]pyridine and quinoxaline 2-substituted derivatives.
Farmaco (Societa chimica italiana : 1989) (1993-09-01)
S Piras, M Loriga, G Paglietti, F Sparatore, M P Demontis, M V Varoni, M C Fattaccio, V Anania
RESUMEN
Twenty compounds possessing benzimidazole, imidazo[4,5-b]pyridine and quinoxaline structure bearing either a substituted arylmethylmercapto- or an arylmethylsulfinyl group in position 2 were prepared in order to evaluate an antiulcer and gastroprotective activity in rat pylorus ligature, in comparison with omeprazole at the dose of 50 mg/kg after i.m. administration. One third of these compounds showed a moderate activity, being about half potent as omeprazole.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Azabenzimidazole, 99%