Merck
Interaction between phencyclidine and its pyrolysis product, 1-phenylcyclohexene.
Pharmacology, biochemistry, and behavior (1988-08-01)
A K Chaturvedi, D J Kuntz
RESUMEN
The interaction between phencyclidine (PCP) and its pyrolysis product, 1-phenylcyclohexene (PC), at metabolic level was evaluated in Swiss male mice (21-24 g). PC (1.1, 2.2 and 4.4 mmol/kg/day for 4 days, IP, in corn oil) treatment to mice induced the in vitro metabolism (p less than 0.05) of amidopyrine (17%), aniline (12%), phenacetin (62-100%), pentobarbital (20-26%), PCP (25-80%) and benzo[a]pyrene (81-147%) in the 9000 g liver fraction and the hepatic microsomal contents of cytochrome P-450 (18-42%). The induction of the mixed function oxygenase (MFO) system was consistent with the decreases in the concentrations of IP administered pentobarbital (0.27 mmol/kg, in saline) and PCP (16.4, 32.8 and 65.6 mumol/kg, in saline) in the serum, brain, liver and kidneys of PC pretreated mice. At 1 hr after the above doses of PC, the in vitro metabolism of amidopyrine, aniline, or phenacetin was not inhibited. However, the biotransformation of benzo[a]pyrene was inhibited by 33 to 45%. Though PC after a single dose did not alter the tissue concentrations of PCP, it increased the pentobarbital concentrations in the tissues studied (p less than 0.05). These results indicate that PC has a potential to induce the MFO system after the 4-day treatment. This property of PC plays an important role in the reduction of the action of PCP by enhancing its metabolism, thereby decreasing its tissue levels.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Phenyl-1-cyclohexene, 95%