- Antagonism of ethanol intoxication in rats by inhibitors of phenylethanolamine N-methyltransferase.
Antagonism of ethanol intoxication in rats by inhibitors of phenylethanolamine N-methyltransferase.
Alcoholism, clinical and experimental research (1990-02-01)
I N Mefford, R G Lister, M Ota, M Linnoila
PMID2178473
RESUMEN
The probable involvement of brain epinephrine in the expression of the acute sedative and intoxicating effects of ethanol and pentobarbital is demonstrated. Two selective inhibitors of phenylethanolamine N-methyltransferase (PNMT), LY134046 and LY78335, proved to be potent and long-lasting antagonists of ethanol intoxication in rats. Acute antagonism of pentobarbital-induced intoxication was observed with LY134046. The present results are compatible with a role for central epinephrine synthesis in ethanol and pentobarbital-induced sedation and intoxication in rats.