Merck
Field nanoemitter: one-dimension Al4C3 ceramics.
Nanoscale (2011-06-21)
Y Sun, H Cui, L Gong, Jian Chen, P K Shen, C X Wang
RESUMEN
As a kind of ionic (or salt-like) carbide, Al(4)C(3) hardly any active functions have been found except for structure material purposes. However, considering the unique characteristic features of its crystal structure, we think Al(4)C(3) in fact might have huge potential for exhibiting active functionality on field-emission application. Herein, we report for the first time the catalyst-free synthesis and excellent field emission properties of Al(4)C(3) one-dimension (1-D) nanostructures. The 1-D nanostructures acting as cold electron emitters display excellent field emission performance with the turn-on field as low as 1.4-2.0 V μm(-1) and the threshold field down to 4.2-4.4 V μm(-1). Such emitters are technologically useful, because they can be easily fabricated on large substrates, and the synthesis process is simple and broadly applicable. The findings conceptually provide new opportunities for the application of Al(4)C(3) ceramic material in vacuum microelectronic devices.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Aluminum carbide, powder, ~325 mesh, 95%