Merck
Lactones and quinones from the tubers of Sinningia aggregata.
Journal of natural products (2010-08-06)
Maria Helena Verdan, Andersson Barison, Eduardo Lemos de Sá, Marcos J Salvador, Clarisse B Poliquesi, Marcos N Eberlin, Maria Elida A Stefanello
RESUMEN
Three new aromatic epsilon-lactones, aggregatins A (1), B (2), and C (3), a new naphthoquinone derivative, aggregatin D (4), and three known anthraquinones, 2-methylanthraquinone, 7-methoxy-2-methylanthraquinone, and 7-hydroxy-2-methylanthraquinone, were isolated from the tubers of Sinningia aggregata (Gesneriaceae). Compounds 1 and 4 and the anthraquinones showed marginal antimicrobial activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Methylanthraquinone, technical, ≥95% (HPLC)