Merck
  • Statins, antihypertensive treatment, and blood pressure control in clinic and over 24 hours: evidence from PHYLLIS randomised double blind trial.
Statins, antihypertensive treatment, and blood pressure control in clinic and over 24 hours: evidence from PHYLLIS randomised double blind trial.
BMJ (Clinical research ed.) (2010-03-27)
Giuseppe Mancia, Gianfranco Parati, Miriam Revera, Grzegorz Bilo, Andrea Giuliano, Fabrizio Veglia, Gaetano Crepaldi, Alberto Zanchetti
RESUMEN
To investigate the possibility that statins reduce blood pressure as well as cholesterol concentrations through clinic and 24 hour ambulatory blood pressure monitoring. Randomised placebo controlled double blind trial. 13 hospitals in Italy 508 patients with mild hypertension and hypercholesterolaemia, aged 45 to 70 years. Participants were randomised to antihypertensive treatment (hydrochlorothiazide 25 mg once daily or fosinopril 20 mg once daily) with or without the addition of a statin (pravastatin 40 mg once daily). Main outcome measures Clinic and ambulatory blood pressure measured every year throughout an average 2.6 year treatment period. Both the group receiving antihypertensive treatment without pravastatin (n=254) (with little change in total cholesterol) and the group receiving antihypertensive treatment with pravastatin (n=253) (with marked and sustained reduction in total cholesterol and low density lipoprotein cholesterol) had a clear cut sustained reduction in clinic measured systolic and diastolic blood pressure as well as in 24 hour, and day and night, systolic and diastolic blood pressure. Pravastatin performed slightly worse than placebo, and between group differences did not exceed 1.9 (95% confidence interval -0.6 to 4.3, P=0.13) mm Hg throughout the treatment period. This was also the case when participants who remained on monotherapy with hydrochlorothiazide or fosinopril throughout the study were considered separately. Administration of a statin in hypertensive patients in whom blood pressure is effectively reduced by concomitant antihypertensive treatment does not have an additional blood pressure lowering effect. Trial registration BRISQUI_*IV_2004_001 (registered at Osservatorio Nazionale sulla Sperimentazione Clinica dei Medicinali-National Monitoring Centre on Clinical Research with Medicines).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fosinopril sodium, ≥98% (HPLC), powder