Merck
  • Inhibition of monoamine oxidases desensitizes 5-HT1A autoreceptors and allows nicotine to induce a neurochemical and behavioral sensitization.
Inhibition of monoamine oxidases desensitizes 5-HT1A autoreceptors and allows nicotine to induce a neurochemical and behavioral sensitization.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-01-30)
Christophe Lanteri, Sandra Jimena Hernández Vallejo, Lucas Salomon, Emilie Lucie Doucet, Gérard Godeheu, Yvette Torrens, Vanessa Houades, Jean-Pol Tassin
RESUMEN
Although nicotine is generally considered to be the main compound responsible for addictive properties of tobacco, experimental data indicate that nicotine does not exhibit all the characteristics of other substances of abuse. We recently showed that a pretreatment with mixed irreversible monoamine oxidases inhibitors (MAOIs), such as tranylcypromine, triggers a locomotor response to nicotine in mice and allows maintenance of behavioral sensitization to nicotine in rats. Moreover, we showed by microdialysis in mice that behavioral sensitization induced by compounds belonging to main groups of drugs of abuse, such as amphetamine, cocaine, morphine, or alcohol, was underlain by sensitization of noradrenergic and serotonergic neurons. Here, this neurochemical sensitization was tested after nicotine, tranylcypromine, or a mixture of both compounds. Data indicate that, whereas neither repeated nicotine nor repeated tranylcypromine alone has any effect by itself, a repeated treatment with a mixture of nicotine and tranylcypromine induces both behavioral sensitization and sensitization of noradrenergic and serotonergic neurons. The development of neurochemical and behavioral sensitizations is blocked by prazosin and SR46349B [(1Z,2E)-1-(2-fluoro-phenyl)-3-(4-hydroxyphenyl)-prop-2-en-one-O-(2-dimethylamino-ethyl)-oxime hemifumarate], two antagonists of alpha1b-adrenergic and 5-HT(2A) receptors, respectively, but not by SCH23390 [R(+)-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride], a D(1) receptor antagonist. Finally, we found that pretreatments with WAY 100635 [N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-(2-pyridinyl)cyclo-hexane carboxamide trihydrochloride], a 5-HT(1A) receptor antagonist, can also induce a behavioral and neurochemical sensitization to repeated nicotine. Complementary experiments with 8-OHDPAT (8-hydroxy-dipropylamino-tetralin), a 5-HT(1A) receptor agonist, and analysis of 5-HT(1A) receptors expression in the dorsal raphe nucleus after a tranylcypromine injection indicate that MAOIs contained in tobacco desensitize 5-HT(1A) autoreceptors to trigger the strong addictive properties of tobacco.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
SR-46349 hemifumarate salt, ≥98% (HPLC)