Merck
Hydrolysis kinetics of 1,3-benzoxazine-2,4-dione (a potential salicylamide prodrug) and various N-substituted derivatives.
Acta pharmaceutica Nordica (1991-01-01)
A H Kahns, H Bundgaard
RESUMEN
The kinetics and mechanism of hydrolysis of 1,3-benzoxazine-2,4-dione and its N-methyl and N-benzoyl derivatives were studied in aqueous solution to provide basic information on the reactivity of the benzoxazinedione structure and to assess the potential of unsubstituted 1,3-benzoxazine-2,4-dione as a prodrug for salicylamide. The compounds were found to hydrolyze quantitatively to the parent salicylamide. The pH-rate profiles obtained at pH 1-11 were accounted for by a spontaneous or water-catalyzed reaction which predominated at pH 1-4 and a hydroxide ion-catalyzed reaction. The rates of hydrolysis were catalyzed slightly in the presence of human plasma and rat liver homogenate, the exception being the N-benzoyl derivative which was hydrolyzed very fast in plasma solutions to N-benzoylsalicylamide. The aqueous solubility and lipophilicity characteristics of 1,3-benzoxazine-2,4-dione were determined. The results obtained suggest that the latter may function as a prodrug for salicylamide with the potential of depressing the extensive first-pass metabolism of salicylamide following oral or rectal administration.