Merck
A saturated N-acylethanolamine other than N-palmitoyl ethanolamine with anti-inflammatory properties: a neglected story...
Journal of neuroendocrinology (2008-05-09)
M Dalle Carbonare, E Del Giudice, A Stecca, D Colavito, M Fabris, A D'Arrigo, D Bernardini, M Dam, A Leon
RESUMEN
N-acylethanolamines, which include the endocannabinoid anandamide and the cannabinoid receptor-inactive saturated compounds N-palmitoyl ethanolamine and N-stearoyl ethanolamine, are ethanolamines of long-chain fatty acids degraded by fatty acid amide hydrolase (FAAH) known to accumulate in degenerating tissues and cells. Whilst much evidence supports a protective anti-inflammatory role of both anandamide and N-palmitoyl ethanolamine, very little information is available with regard to the bioactivity of N-stearoyl ethanolamine. Employing a murine model of passive IgE-induced cutaneous anaphylaxis, we have found that N-stearoyl ethanolamine is endowed with marked anti-inflammatory properties in vivo, supporting the hypothesis that endogenous N-stearoyl ethanolamine is, in analogy to N-palmitoyl ethanolamine, a bioactive signalling lipid capable of downregulating allergic inflammation in the skin. This effect, although mimicked by synthetic, non-selective, CB(1)/CB(2) receptor agonists, such as WIN55, 212-2, was not sensitive to CB(1) or CB(2) receptor antagonists, but rather was fully reversed by capsazepine, a competitive antagonist of the TRPV1 receptor. Moreover, CB(1) receptor antagonists, although effective in antagonising the WIN55,212-2-induced hypothermia, did not reduce the anti-inflammatory effect of WIN55,212-2, whilst CB(2) receptor antagonists, per se inactive, potentiated the WIN55,212-2 effect, suggesting an involvement of non-CB(1)/CB(2) receptors in the anti-inflammatory action of WIN55,212-2. All this, together with demonstration of FAAH as a major regulator of the in vivo concentrations of saturated N-stearoyl ethanolamine, in addition to N-palmitoyl ethanolamine, raise the speculation that pharmacological treatments with saturated N-acylethanolamines such as N-stearoyl ethanolamine, or alternatively FAAH inhibitors able to increase their local concentration, rather than selective CB receptor agonists, might be of promising therapeutic benefit in reducing allergic inflammation in the skin.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Stearoyl ethanolamide, ≥98%, crystalline