- The ansacarbamitocins: polar ansamitocin derivatives.
The ansacarbamitocins: polar ansamitocin derivatives.
Journal of natural products (2007-09-26)
Carl E Snipes, Dennis O Duebelbeis, Monica Olson, Donald R Hahn, William H Dent, Jeffrey R Gilbert, Todd L Werk, George E Davis, Rebecca Lee-Lu, Paul R Graupner
PMID17892263
RESUMEN
The ansacarbamitocins are a new family of maytansinoids that are unusually substituted with a glucose subunit and two carbamate functional groups and exhibit modest activity against some agricultural fungal disease organisms. Ansacarbamitocins A-F ( 1- 6) all consist of the same macrocyclic core as the ansamitocins, with variation occurring on the glucose unit, while ansacarbamitocins A1 and B1 ( 7, 8) additionally lack the epoxide group on C-4 and C-5.