Merck
Calcium chloride and calcium bromide aqueous solutions of technical and analytical grade in Lemna bioassay.
Chemosphere (2000-11-01)
M Vujević, Z Vidaković-Cifrek, M Tkalec, M Tomíc, I Regula
RESUMEN
Saturated water solutions of calcium chloride, calcium bromide and their 1:1 mixture are commonly used as "high density brines" for pressure control in oil wells. To compare the effect of these chemicals of technical grade with the effect of the chemicals of analytical grade the Lemna test was used. The multiplication rate, fresh weight, dry to fresh weight ratio, area covered by plants and chlorophyll content were measured as toxicity parameters. The concentrations of tested chemicals were 0.025, 0.05. 0.075 and 0.1 mol dm(-3). Generally, the chemicals of both technical and analytical grade in concentrations of 0.025 mol dm(-3) stimulated the Lemna minor growth, while tested chemicals in concentrations of 0.05 mol dm(-3) did not affect the growth significantly. The exceptions were results obtained by measuring fresh weight. Most of tested chemicals in concentrations of 0.075 mol dm(-3) and all chemicals in concentrations of 0.1 mol dm(-3) reduced the growth. No major differences between effects of tested chemicals of technical and analytical grade on plant growth were observed, except that tested chemicals of analytical grade in concentrations of 0.1 mol dm(-3) increased dry to fresh weight ratio much stronger than chemicals of technical grade. All tested chemicals in all concentrations increased chlorophyll content. After treatment with chemicals of analytical grade much higher increase of chlorophyll a concentration in comparison to increase of chlorophyll b was noticed, while chemicals of technical grade caused more prominent increase of chlorophyll b.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Calcium bromide, AnhydroBeads, −10 mesh, 99.98% trace metals basis